Assay Details
Gene Symbol: MPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343318.1 | NP_001330561.1 | 3 - 4 | 486 | 126 | |
NM_001343319.1 | NP_001330562.1 | 4 - 5 | 698 | 126 | ||
NM_001343320.1 | NP_001330559.1 | 3 - 4 | 364 | 126 | ||
NM_001343321.1 | NP_001330560.1 | 4 - 5 | 698 | 126 | ||
NM_121422.4 | NP_568295.2 | 4 - 5 | 836 | 126 | ||
GenBank mRNA | AK176026.1 | - | 4 - 5 | 596 | 126 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
