Assay Details
Gene Symbol: MPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343318.1 | NP_001330561.1 | 6 - 7 | 760 | 148 | |
NM_001343319.1 | NP_001330562.1 | 7 - 8 | 972 | 148 | ||
NM_001343320.1 | NP_001330559.1 | 6 - 7 | 638 | 148 | ||
NM_121422.4 | NP_568295.2 | 7 - 8 | 1110 | 148 | ||
GenBank mRNA | AK176026.1 | - | 7 - 8 | 870 | 148 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
