Assay Details
Gene Symbol: HEMG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203378.1 | NP_001190307.1 | 9 - 10 | 588 | 98 | |
NM_001343324.1 | NP_001331496.1 | 9 - 10 | 992 | 98 | ||
NM_001343325.1 | NP_001331495.1 | 10 - 11 | 883 | 98 | ||
NM_001343326.1 | NP_001331494.1 | 10 - 11 | 694 | 98 | ||
NM_121426.5 | NP_196926.2 | 10 - 11 | 657 | 98 | ||
GenBank mRNA | AY075665.1 | - | 10 - 11 | 632 | 98 | |
AY101523.1 | - | 10 - 11 | 599 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
