Assay Details
Gene Symbol: HEMG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203378.1 | NP_001190307.1 | 13 - 14 | 1107 | 120 | |
NM_001343324.1 | NP_001331496.1 | 13 - 14 | 1511 | 120 | ||
NM_001343325.1 | NP_001331495.1 | 14 - 15 | 1402 | 120 | ||
NM_001343326.1 | NP_001331494.1 | 14 - 15 | 1213 | 120 | ||
NM_121426.5 | NP_196926.2 | 14 - 15 | 1176 | 120 | ||
GenBank mRNA | AY075665.1 | - | 14 - 15 | 1151 | 120 | |
AY101523.1 | - | 14 - 15 | 1118 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
