Assay Details
Gene Symbol: HEMG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203378.1 | NP_001190307.1 | 15 - 16 | 1280 | 127 | |
NM_001343324.1 | NP_001331496.1 | 15 - 16 | 1684 | 127 | ||
NM_001343325.1 | NP_001331495.1 | 16 - 17 | 1575 | 127 | ||
NM_001343326.1 | NP_001331494.1 | 16 - 17 | 1386 | 127 | ||
NM_121426.5 | NP_196926.2 | 16 - 17 | 1349 | 127 | ||
GenBank mRNA | AY075665.1 | - | 16 - 17 | 1324 | 127 | |
AY101523.1 | - | 16 - 17 | 1291 | 127 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
