Assay Details
Gene Symbol: HEMG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203378.1 | NP_001190307.1 | 2 - 3 | 162 | 99 | |
NM_001343325.1 | NP_001331495.1 | 2 - 3 | 367 | 99 | ||
NM_121426.5 | NP_196926.2 | 2 - 3 | 141 | 99 | ||
GenBank mRNA | AY075665.1 | - | 2 - 3 | 116 | 99 | |
AY101523.1 | - | 2 - 3 | 83 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
