Assay Details
Gene Symbol: HEMG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203378.1 | NP_001190307.1 | 3 - 4 | 290 | 111 | |
NM_001343324.1 | NP_001331496.1 | 2 - 3 | 604 | 111 | ||
NM_001343325.1 | NP_001331495.1 | 3 - 4 | 495 | 111 | ||
NM_001343326.1 | NP_001331494.1 | 3 - 4 | 306 | 111 | ||
NM_121426.5 | NP_196926.2 | 3 - 4 | 269 | 111 | ||
GenBank mRNA | AY075665.1 | - | 3 - 4 | 244 | 111 | |
AY101523.1 | - | 3 - 4 | 211 | 111 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001343324.1, NM_001343326.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
