Assay Details
Gene Symbol: HEMG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343324.1 | NP_001331496.1 | 6 - 7 | 860 | 133 | |
NM_001343325.1 | NP_001331495.1 | 7 - 8 | 751 | 133 | ||
NM_001343326.1 | NP_001331494.1 | 7 - 8 | 562 | 133 | ||
NM_121426.5 | NP_196926.2 | 7 - 8 | 525 | 133 | ||
GenBank mRNA | AY075665.1 | - | 7 - 8 | 500 | 133 | |
AY101523.1 | - | 7 - 8 | 467 | 133 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
