Assay Details
Gene Symbol: CA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036806.2 | NP_001031883.1 | 2 - 3 | 604 | 70 | |
NM_001036807.1 | NP_001031884.1 | 2 - 3 | 488 | 70 | ||
NM_001085114.1 | NP_001078583.1 | 2 - 3 | 488 | 70 | ||
NM_001343362.1 | NP_001330238.1 | 1 - 2 | 291 | 70 | ||
NM_121478.4 | NP_568303.2 | 2 - 3 | 526 | 70 | ||
GenBank mRNA | AF324712.2 | - | 1 - 2 | 83 | 70 | |
AF326863.1 | - | 1 - 2 | 83 | 70 | ||
AF428428.1 | - | 1 - 2 | 124 | 70 | ||
BT000652.1 | - | 1 - 2 | 81 | 70 | ||
BT000663.1 | - | 1 - 2 | 85 | 70 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001343362.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
