Assay Details
| Gene Symbol: BCCP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121557.4 | NP_568316.1 | 6 - 7 | 1136 | 62 | |
| GenBank mRNA | AF223948.1 | - | 6 - 7 | 837 | 62 | |
| AY086528.1 | - | 6 - 7 | 927 | 62 | ||
More Information
Set Membership: |
|

BCCP2
Genomic Map