Assay Details
| Gene Symbol: PAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121598.4 | NP_197097.1 | 5 - 6 | 880 | 98 | |
| GenBank mRNA | AF069528.1 | - | 5 - 6 | 813 | 98 | |
| AY087917.1 | - | 5 - 6 | 845 | 98 | ||
More Information
Set Membership: |
|

PAM1
Genomic Map