Assay Details
Gene Symbol: SYP21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121689.4 | NP_197185.1 | 6 - 7 | 914 | 112 | |
GenBank mRNA | AF411797.1 | - | 6 - 7 | 832 | 112 | |
BT025036.1 | - | 6 - 7 | 771 | 112 | ||
L41651.1 | - | 6 - 7 | 875 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
