Assay Details
| Gene Symbol: XPO1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203395.1 | NP_001190324.1 | 20 - 21 | 2132 | 94 | |
| NM_121708.3 | NP_197204.1 | 21 - 22 | 2269 | 94 | ||
| GenBank mRNA | AY056126.1 | - | 21 - 22 | 2165 | 94 | |
| AY096633.1 | - | 21 - 22 | 1997 | 94 | ||
| Y18469.1 | - | 2092 | 94 | |||
More Information
Set Membership: |
|

XPO1A
Genomic Map