Assay Details
| Gene Symbol: AT5G17200 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121726.1 | NP_197222.1 | 1 - 2 | 166 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT5G17200| Gene Symbol: AT5G17200 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121726.1 | NP_197222.1 | 1 - 2 | 166 | 77 | |
Set Membership: |
|