Assay Details
Gene Symbol: PC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343504.1 | NP_001318585.1 | 1 | 244 | 95 | |
GenBank mRNA | DQ446958.1 | - | 1 | 155 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
