Assay Details
| Gene Symbol: CBR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121783.5 | NP_197279.1 | 1 - 2 | 429 | 113 | |
| GenBank mRNA | AB007799.1 | - | 1 - 2 | 197 | 113 | |
| AK316787.1 | - | 1 - 2 | 140 | 113 | ||
| AY085728.1 | - | 1 - 2 | 268 | 113 | ||
More Information
Set Membership: |
|

CBR
Genomic Map