Assay Details
| Gene Symbol: DAR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121795.4 | NP_197291.2 | 3 - 4 | 1988 | 157 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DAR4| Gene Symbol: DAR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121795.4 | NP_197291.2 | 3 - 4 | 1988 | 157 | |
Set Membership: |
|