Assay Details
| Gene Symbol: GDH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121822.4 | NP_197318.1 | 1 - 2 | 241 | 106 | |
| GenBank mRNA | BT025561.1 | - | 1 - 2 | 104 | 106 | |
| U37771.1 | - | 1 - 2 | 205 | 106 | ||
| U53527.1 | - | 1 - 2 | 203 | 106 | ||
More Information
Set Membership: |
|

GDH1
Genomic Map