Assay Details
| Gene Symbol: GDH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121822.4 | NP_197318.1 | 6 - 7 | 1030 | 78 | |
| GenBank mRNA | BT025561.1 | - | 6 - 7 | 893 | 78 | |
| U37771.1 | - | 6 - 7 | 994 | 78 | ||
| U53527.1 | - | 6 - 7 | 992 | 78 | ||
More Information
Set Membership: |
|

GDH1
Genomic Map