Assay Details
| Gene Symbol: GDH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121822.4 | NP_197318.1 | 8 - 9 | 1191 | 96 | |
| GenBank mRNA | BT025561.1 | - | 8 - 9 | 1054 | 96 | |
| U37771.1 | - | 8 - 9 | 1155 | 96 | ||
| U53527.1 | - | 8 - 9 | 1153 | 96 | ||
More Information
Set Membership: |
|

GDH1
Genomic Map