Assay Details
| Gene Symbol: AT5G18180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121823.2 | NP_197319.1 | 2 - 3 | 447 | 118 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT5G18180| Gene Symbol: AT5G18180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121823.2 | NP_197319.1 | 2 - 3 | 447 | 118 | |
Set Membership: |
|