Assay Details
| Gene Symbol: AT5G19250 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121930.5 | NP_197426.1 | 1 - 2 | 241 | 78 | |
| GenBank mRNA | BT014784.1 | - | 1 - 2 | 95 | 78 | |
| BT015011.1 | - | 1 - 2 | 85 | 78 | ||
More Information
Set Membership: |
|

AT5G19250
Genomic Map