Assay Details
| Gene Symbol: CHR23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343595.1 | NP_001330405.1 | 4 - 5 | 1031 | 83 | |
| NM_121936.3 | NP_197432.2 | 8 - 9 | 1758 | 83 | ||
More Information
Set Membership: |
|

CHR23
Genomic Map