Assay Details
| Gene Symbol: ASP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121960.5 | NP_197456.1 | 1 - 2 | 285 | 86 | |
| GenBank mRNA | BT025972.1 | - | 1 - 2 | 55 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ASP2| Gene Symbol: ASP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121960.5 | NP_197456.1 | 1 - 2 | 285 | 86 | |
| GenBank mRNA | BT025972.1 | - | 1 - 2 | 55 | 86 | |
Set Membership: |
|