Assay Details
Gene Symbol: ROPGEF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343611.1 | NP_001318601.1 | 4 - 5 | 584 | 102 | |
NM_001343612.1 | NP_001331396.1 | 3 - 4 | 779 | 102 | ||
NM_001343614.1 | NP_001331397.1 | 3 - 4 | 779 | 102 | ||
NM_121961.2 | NP_197457.1 | 3 - 4 | 398 | 102 | ||
GenBank mRNA | DQ459196.1 | - | 3 - 4 | 398 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
