Assay Details
Gene Symbol: ROPGEF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343611.1 | NP_001318601.1 | 5 - 6 | 657 | 86 | |
NM_001343612.1 | NP_001331396.1 | 4 - 5 | 852 | 86 | ||
NM_001343613.1 | NP_001331395.1 | 4 - 5 | 868 | 86 | ||
NM_001343614.1 | NP_001331397.1 | 4 - 5 | 852 | 86 | ||
NM_121961.2 | NP_197457.1 | 4 - 5 | 471 | 86 | ||
GenBank mRNA | DQ459196.1 | - | 4 - 5 | 471 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
