Assay Details
Gene Symbol: SKdZeta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128652.4 | NP_180655.1 | 6 - 7 | 773 | 104 | |
GenBank mRNA | AY064020.1 | - | 6 - 7 | 723 | 104 | |
AY094423.1 | - | 6 - 7 | 749 | 104 | ||
AY096698.1 | - | 6 - 7 | 670 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
