Assay Details
Gene Symbol: MYB14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128674.4 | NP_180676.1 | 1 - 2 | 338 | 119 | |
GenBank mRNA | AY519575.1 | - | 1 - 2 | 133 | 119 | |
BT024853.1 | - | 1 - 2 | 133 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
