Assay Details
| Gene Symbol: GPX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128714.4 | NP_180715.1 | 2 - 3 | 273 | 116 | |
| GenBank mRNA | AY044330.1 | - | 2 - 3 | 125 | 116 | |
| AY058187.1 | - | 2 - 3 | 228 | 116 | ||
| AY086518.1 | - | 2 - 3 | 212 | 116 | ||
| AY098982.1 | - | 2 - 3 | 125 | 116 | ||
| U94495.1 | - | 2 - 3 | 150 | 116 | ||
More Information
Set Membership: |
|

GPX2
Genomic Map