Assay Details
Gene Symbol: CCT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336383.1 | NP_001323485.1 | 2 - 3 | 983 | 115 | |
NM_128785.3 | NP_180785.1 | 3 - 4 | 581 | 115 | ||
GenBank mRNA | AY136393.1 | - | 3 - 4 | 514 | 115 | |
BT000207.1 | - | 3 - 4 | 393 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
