Assay Details
Gene Symbol: CCT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336383.1 | NP_001323485.1 | 4 - 5 | 1180 | 106 | |
NM_128785.3 | NP_180785.1 | 5 - 6 | 778 | 106 | ||
GenBank mRNA | AY136393.1 | - | 5 - 6 | 711 | 106 | |
BT000207.1 | - | 5 - 6 | 590 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
