Assay Details
| Gene Symbol: BAM6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128788.3 | NP_180788.2 | 2 - 3 | 741 | 143 | |
| GenBank mRNA | AY136463.1 | - | 2 - 3 | 668 | 143 | |
More Information
Set Membership: |
|

BAM6
Genomic Map