Assay Details
| Gene Symbol: BAM6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128788.3 | NP_180788.2 | 6 - 7 | 1620 | 135 | |
| GenBank mRNA | AY136463.1 | - | 6 - 7 | 1547 | 135 | |
More Information
Set Membership: |
|

BAM6
Genomic Map