Assay Details
| Gene Symbol: EMB2795 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128818.3 | NP_180818.2 | 4 - 5 | 687 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EMB2795| Gene Symbol: EMB2795 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128818.3 | NP_180818.2 | 4 - 5 | 687 | 85 | |
Set Membership: |
|