Assay Details
Gene Symbol: LUH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036389.4 | NP_001031466.2 | 5 - 6 | 519 | 82 | |
NM_001202730.1 | NP_001189659.1 | 5 - 6 | 507 | 82 | ||
NM_128829.4 | NP_565749.1 | 5 - 6 | 519 | 82 | ||
NM_179861.3 | NP_850192.1 | 5 - 6 | 465 | 82 | ||
NM_179862.2 | NP_850193.1 | 4 - 5 | 594 | 82 | ||
NM_179863.3 | NP_850194.1 | 5 - 6 | 561 | 82 | ||
NM_179864.3 | NP_850195.2 | 5 - 6 | 410 | 82 | ||
GenBank mRNA | AF367306.1 | - | 5 - 6 | 507 | 82 | |
BT002219.1 | - | 3 - 4 | 240 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
