Assay Details
Gene Symbol: BGAL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202731.1 | NP_001189660.1 | 13 - 14 | 1676 | 77 | |
NM_128841.5 | NP_565755.1 | 13 - 14 | 1746 | 77 | ||
GenBank mRNA | AJ270305.1 | - | 13 - 14 | 1650 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
