Assay Details
| Gene Symbol: BGLU33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128846.4 | NP_180845.2 | 9 - 10 | 1355 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BGLU33| Gene Symbol: BGLU33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128846.4 | NP_180845.2 | 9 - 10 | 1355 | 85 | |
Set Membership: |
|