Assay Details
| Gene Symbol: GLTP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128907.5 | NP_565766.1 | 2 - 3 | 541 | 103 | |
| NM_201859.4 | NP_973588.1 | 2 - 3 | 522 | 103 | ||
| GenBank mRNA | AK317264.1 | - | 2 - 3 | 204 | 103 | |
| BT004207.1 | - | 2 - 3 | 209 | 103 | ||
| BT005674.1 | - | 1 - 2 | 116 | 103 | ||
More Information
Set Membership: |
|

GLTP1
Genomic Map