Assay Details
| Gene Symbol: GLTP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128907.5 | NP_565766.1 | 4 - 5 | 726 | 69 | |
| NM_201859.4 | NP_973588.1 | 4 - 5 | 707 | 69 | ||
| GenBank mRNA | AK317264.1 | - | 4 - 5 | 389 | 69 | |
| AY085884.1 | - | 4 - 5 | 488 | 69 | ||
| BT004207.1 | - | 4 - 5 | 394 | 69 | ||
| BT005674.1 | - | 3 - 4 | 301 | 69 | ||
More Information
Set Membership: |
|

GLTP1
Genomic Map