Assay Details
| Gene Symbol: GLTP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128907.5 | NP_565766.1 | 5 - 6 | 870 | 60 | |
| NM_201859.4 | NP_973588.1 | 5 - 6 | 851 | 60 | ||
| GenBank mRNA | AK317264.1 | - | 5 - 6 | 533 | 60 | |
| AY085884.1 | - | 5 - 6 | 632 | 60 | ||
| BT004207.1 | - | 5 - 6 | 538 | 60 | ||
| BT005674.1 | - | 4 - 5 | 445 | 60 | ||
More Information
Set Membership: |
|

GLTP1
Genomic Map