Assay Details
| Gene Symbol: CPL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336459.1 | NP_001325202.1 | 3 - 4 | 1396 | 128 | |
| NM_128914.3 | NP_180912.2 | 4 - 5 | 1271 | 128 | ||
More Information
Set Membership: |
|

CPL3
Genomic Map