Assay Details
| Gene Symbol: SWI3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128921.6 | NP_180919.2 | 2 - 3 | 616 | 91 | |
| GenBank mRNA | BT004085.1 | - | 2 - 3 | 598 | 91 | |
| BT005094.1 | - | 2 - 3 | 565 | 91 | ||
More Information
Set Membership: |
|

SWI3B
Genomic Map