Assay Details
| Gene Symbol: ETT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128946.4 | NP_180942.1 | 1 - 2 | 685 | 84 | |
| GenBank mRNA | AF336917.1 | - | 1 - 2 | 320 | 84 | |
| AF360313.1 | - | 1 - 2 | 418 | 84 | ||
| AY056365.1 | - | 1 - 2 | 320 | 84 | ||
More Information
Set Membership: |
|

ETT
Genomic Map