Assay Details
| Gene Symbol: ETT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336475.1 | NP_001324363.1 | 2 - 3 | 687 | 72 | |
| NM_128946.4 | NP_180942.1 | 3 - 4 | 931 | 72 | ||
| GenBank mRNA | AF336917.1 | - | 3 - 4 | 566 | 72 | |
| AF360313.1 | - | 3 - 4 | 664 | 72 | ||
| AY056365.1 | - | 3 - 4 | 566 | 72 | ||
More Information
Set Membership: |
|

ETT
Genomic Map