Assay Details
| Gene Symbol: HB-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336476.1 | NP_001324695.1 | 2 - 3 | 1133 | 78 | |
| NM_128948.4 | NP_180944.2 | 2 - 3 | 1151 | 78 | ||
| GenBank mRNA | AY251401.1 | - | 2 - 3 | 1099 | 78 | |
More Information
Set Membership: |
|

HB-3
Genomic Map