Assay Details
Gene Symbol: SYP81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333499.1 | NP_001323219.1 | 4 - 5 | 1018 | 97 | |
NM_104052.3 | NP_564597.1 | 5 - 6 | 871 | 97 | ||
GenBank mRNA | BT002917.1 | - | 5 - 6 | 840 | 97 | |
BT004352.1 | - | 5 - 6 | 651 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
