Assay Details
| Gene Symbol: MBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104085.4 | NP_175616.1 | 2 - 3 | 288 | 109 | |
| GenBank mRNA | AY090380.1 | - | 1 - 2 | 207 | 109 | |
More Information
Set Membership: |
|

MBP1
Genomic Map