Assay Details
| Gene Symbol: PIP1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084854.2 | NP_001078323.1 | 4 - 5 | 1020 | 86 | |
| NM_116008.4 | NP_191702.1 | 3 - 4 | 1177 | 86 | ||
| GenBank mRNA | AK229222.1 | - | 3 - 4 | 830 | 86 | |
| AY058113.1 | - | 3 - 4 | 830 | 86 | ||
| AY097398.1 | - | 3 - 4 | 763 | 86 | ||
More Information
Set Membership: |
|

PIP1A
Genomic Map