Assay Details
Gene Symbol: DL1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340126.1 | NP_001325984.1 | 11 - 12 | 1194 | 92 | |
NM_001340127.1 | NP_001325985.1 | 11 - 12 | 1194 | 92 | ||
NM_116041.3 | NP_191735.2 | 11 - 12 | 1194 | 92 | ||
GenBank mRNA | AY189279.1 | - | 11 - 12 | 1118 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
