Assay Details
Gene Symbol: DL1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340126.1 | NP_001325984.1 | 12 - 13 | 1474 | 98 | |
NM_116041.3 | NP_191735.2 | 12 - 13 | 1474 | 98 | ||
GenBank mRNA | AY189279.1 | - | 12 - 13 | 1398 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
